Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316341.1 | NP_001303270.1 | 12 | 1504 | ||
NM_002816.4 | NP_002807.1 | 10 | 1220 | |||
NM_174871.3 | NP_777360.1 | 9 | 1160 | |||
GenBank mRNA | AB003103.1 | - | 10 | 1155 | ||
AK091198.1 | - | 9 | 1138 | |||
AK222679.1 | - | 10 | 1137 | |||
AK307821.1 | - | 10 | 1170 | |||
BC019062.2 | - | 10 | 1151 | 4111477 | ||
BC065826.1 | - | 12 | 1424 | 30386487 | ||
KU178288.1 | - | 10 | 1112 | |||
KU178289.1 | - | 10 | 1144 |