Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ss18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161370.1 | NP_001154842.1 | 6, 7 | 838 | ||
NM_009280.2 | NP_033306.2 | 7, 8 | 1060 | |||
GenBank mRNA | AK051601.1 | - | 691 | |||
AK083887.1 | - | 7, 8 | 944 | |||
AK166053.1 | - | 7, 8 | 913 | |||
X93357.1 | - | 7, 8 | 1060 |