Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3rf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146299.1 | NP_001139771.1 | 6 | 1408 | ||
NM_172966.3 | NP_766554.2 | 5 | 1312 | |||
GenBank mRNA | AK028153.1 | - | 5 | 808 | ||
AK029168.1 | - | 5 | 1313 | |||
AK033601.1 | - | 6 | 1452 | |||
BC137955.1 | - | 6 | 1361 | 8860974 | ||
BC145060.1 | - | 6 | 1361 | 9053608 |