Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgat4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290801.1 | NP_001277730.1 | 13, 14 | 1706 | ||
NM_173870.3 | NP_776295.1 | 14, 15 | 1785 | |||
GenBank mRNA | AB053217.1 | - | 13, 14 | 1461 | ||
AK052278.1 | - | 11, 12 | 1282 | |||
AK155724.1 | - | 14, 15 | 1787 |