Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175031.3 | NP_778196.2 | 24 | 3065 | ||
GenBank mRNA | AK029842.1 | - | 24 | 2969 | ||
AK047284.1 | - | 13 | 1512 | |||
AK086716.1 | - | 8 | 849 | |||
BC043103.1 | - | 24 | 3011 | 6406485 | ||
BC058698.1 | - | 21 | 2746 | 6831702 | ||
BC061470.1 | - | 23 | 2874 |