Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Muc15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290786.1 | NP_001277715.1 | 2 | 876 | ||
NM_172979.3 | NP_766567.1 | 2 | 876 | |||
GenBank mRNA | AK028832.1 | - | 1 | 71 | ||
AK052840.1 | - | 2 | 763 | |||
AK142590.1 | - | 2 | 763 | |||
AY198336.1 | - | 2 | 794 | |||
AY198337.1 | - | 2 | 761 | |||
BC116796.1 | - | 2 | 755 | 40126115 | ||
BC119176.1 | - | 2 | 755 | 8733925 |