Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ell3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145973.2 | NP_666085.2 | 5, 6 | 614 | ||
GenBank mRNA | AK139676.1 | - | 6, 7 | 1040 | ||
BC045151.1 | - | 5, 6 | 614 | 6313358 |