Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ell3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145973.2 | NP_666085.2 | 10 | 1129 | ||
GenBank mRNA | AK139676.1 | - | 11 | 1555 | ||
BC045151.1 | - | 10 | 1129 | 6313358 |