Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahcy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016661.3 | NP_057870.3 | 9 | 1238 | ||
GenBank mRNA | AK075629.1 | - | 6 | 810 | ||
AK151097.1 | - | 9 | 1198 | |||
AK153421.1 | - | 9 | 1198 | |||
AK169331.1 | - | 9 | 1198 | |||
AK211648.1 | - | 1 | 132 | |||
AK213608.1 | - | 1 | 131 | |||
BC086781.1 | - | 9 | 1167 | 6828058 | ||
BC108367.1 | - | 9 | 1174 | 4457523 | ||
L32836.1 | - | 9 | 1138 |
Gene Symbol: Gm4737 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK154302.1 | - | 1198 | |||
BC015304.1 | - | 1168 | 4241917 |