Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001882.3 | NP_001001882.3 | 10 | 1115 | ||
NM_001166665.1 | NP_001160137.1 | 10 | 1115 | |||
NM_001166666.1 | NP_001160138.1 | 10 | 1115 | |||
NM_001166667.1 | NP_001160139.1 | 10 | 1115 | |||
NM_001166668.1 | NP_001160140.1 | 10 | 1115 | |||
GenBank mRNA | AK142225.1 | - | 10 | 1101 | ||
AK145145.1 | - | 10 | 1087 | |||
AK173097.1 | - | 10 | 1077 | |||
AY481619.1 | - | 10 | 1054 | |||
AY481620.1 | - | 10 | 1054 | |||
AY481621.1 | - | 10 | 1054 | |||
AY481622.1 | - | 10 | 1054 | |||
AY481623.1 | - | 10 | 1054 | |||
BC031201.1 | - | 4 | 280 | |||
BC057352.1 | - | 10 | 1087 | |||
BC105578.1 | - | 10 | 1027 | 30478629 | ||
BC144977.1 | - | 9 | 867 | 9053525 | ||
BC144978.1 | - | 9 | 867 | 9053526 | ||
BC145658.1 | - | 9 | 867 | 40130961 |