Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Luzp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024452.2 | NP_077772.2 | 4 | 1918 | ||
GenBank mRNA | AK131161.1 | - | 1 | 436 | ||
BC137784.1 | - | 2 | 1682 | 8860803 | ||
BC137786.1 | - | 2 | 1682 | 8860805 | ||
BC171934.1 | - | 1 | 1811 | 9054218 | ||
L49344.1 | - | 3 | 1779 |