Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pank4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK050894.1 | - | 16 | 1876 | ||
BC012520.1 | - | 5 | 379 | |||
BC050089.1 | - | 16 | 1874 | 30018732 |