Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golga3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008146.3 | NP_032172.3 | 22 | 4296 | ||
GenBank mRNA | AK032610.1 | - | 6 | 778 | ||
BC043452.1 | - | 14 | 2158 | |||
BC053002.1 | - | 22 | 4296 | 5709074 | ||
D78270.2 | - | 22 | 4102 |