Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTCH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000264.3 | NP_000255.2 | 14 | 2142 | ||
NM_001083602.1 | NP_001077071.1 | 14 | 2250 | |||
NM_001083603.1 | NP_001077072.1 | 14 | 2096 | |||
NM_001083604.1 | NP_001077073.1 | 14 | 1860 | |||
NM_001083605.1 | NP_001077074.1 | 14 | 1843 | |||
NM_001083606.1 | NP_001077075.1 | 14 | 1942 | |||
NM_001083607.1 | NP_001077076.1 | 14 | 2016 | |||
GenBank mRNA | AB209495.1 | - | 12 | 1723 | ||
AB214500.1 | - | 5 | 583 | |||
AB233422.1 | - | 8 | 938 | |||
U43148.1 | - | 14 | 1942 | |||
U59464.1 | - | 14 | 1954 |