Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnft2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109902.1 | NP_001103372.1 | 8 | 1169 | ||
NM_172998.3 | NP_766586.2 | 9 | 1204 | |||
GenBank mRNA | AK046850.1 | - | 9 | 1205 | ||
AK090256.1 | - | 8 | 1265 | |||
AK148930.1 | - | 8 | 1146 | |||
BC057302.1 | - | 8 | 1131 | 6827287 |