Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp664 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081750.1 | NP_001075219.1 | 4 | 1043 | ||
GenBank mRNA | BC096550.1 | - | 1 | 126 | 30550168 |