Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109749.1 | NP_001103219.1 | 16 | 2100 | ||
NM_001109751.1 | NP_001103221.1 | 15 | 2188 | |||
NM_173004.3 | NP_766592.2 | 16 | 2100 | |||
GenBank mRNA | AK036262.1 | - | 16 | 2070 | ||
AK044694.1 | - | 6 | 849 | |||
AK173340.1 | - | 9 | 1219 | |||
BC046975.1 | - | 13 | 1722 | |||
BC115766.1 | - | 16 | 1990 | 40060288 |