Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp384 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252083.1 | NP_001239012.1 | 11 | 1585 | ||
NM_175557.5 | NP_780766.3 | 11 | 1646 | |||
GenBank mRNA | AK031726.1 | - | 4 | 1701 | ||
AK156026.1 | - | 11 | 1587 | |||
BC046410.1 | - | 2 | 177 | |||
BC064013.1 | - | 11 | 1510 | 6515698 | ||
BQ713783.1 | - | 4 | 425 |