Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Letm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173012.3 | NP_766600.2 | 3 | 824 | ||
GenBank mRNA | AK031563.1 | - | 3 | 678 | ||
AK034050.1 | - | 3 | 835 | |||
AK050943.1 | - | 3 | 990 | |||
AK163507.1 | - | 3 | 679 | |||
BC055685.1 | - | 3 | 824 | 6403034 |