Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115130.1 | NP_001108602.1 | 6 | 6875 | ||
NM_172765.3 | NP_766353.2 | 6 | 6821 | |||
GenBank mRNA | AK053995.1 | - | 1 | 2300 | ||
AK153955.1 | - | 1 | 2002 | |||
BC090965.1 | - | 1 | 1482 |