Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163742.1 | NP_001157214.1 | 5 | 411 | ||
NM_001163743.1 | NP_001157215.1 | 5 | 512 | |||
NM_178420.3 | NP_848507.2 | 5 | 500 | |||
GenBank mRNA | AK089260.1 | - | 5 | 504 | ||
AK141296.1 | - | 5 | 481 | |||
AK145780.1 | - | 5 | 521 | |||
AK145988.1 | - | 5 | 501 | |||
AK166689.1 | - | 5 | 338 | |||
AK170088.1 | - | 5 | 455 | |||
BC034204.1 | - | 4 | 294 | |||
BC050054.1 | - | 5 | 407 | 30012053 |