Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177771.5 | NP_808439.2 | 6 | 886 | ||
GenBank mRNA | AK044774.1 | - | 6 | 997 | ||
AK173029.1 | - | 6 | 886 | |||
AK198584.1 | - | 3 | 238 | |||
BC025563.1 | - | 6 | 882 | 5322893 | ||
BC065167.1 | - | 5 | 583 | |||
CB247282.1 | - | 3 | 234 | |||
CK619719.1 | - | 4 | 492 |