Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Catsperb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173023.2 | NP_766611.2 | 10, 11 | 991 | ||
GenBank mRNA | AK029843.1 | - | 10, 11 | 992 | ||
AK030010.1 | - | 10, 11 | 982 | |||
BC132479.1 | - | 10, 11 | 912 | 40130756 | ||
BC145000.1 | - | 10, 11 | 912 | |||
EF199807.1 | - | 9, 10 | 872 |