Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172053.2 | NP_742050.2 | 4 | 899 | ||
GenBank mRNA | AK031314.1 | - | 4 | 865 | ||
AK173338.1 | - | 4 | 835 | |||
BC034739.1 | - | 4 | 912 | 4951547 |