Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lppr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178756.4 | NP_848871.1 | 5 | 723 | ||
GenBank mRNA | AK034251.1 | - | 5 | 818 | ||
AK043762.1 | - | 5 | 625 | |||
AK047614.1 | - | 5 | 653 | |||
AK087439.1 | - | 5 | 874 | |||
AK087514.1 | - | 5 | 724 | |||
AK160897.1 | - | 5 | 850 | |||
AY345342.1 | - | 5 | 723 | |||
BC096762.1 | - | 5 | 699 | 30094664 |