Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tarsl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172310.2 | NP_758514.2 | 3, 4 | 574 | ||
GenBank mRNA | AK040927.1 | - | 3, 4 | 547 | ||
AK041280.1 | - | 3, 4 | 574 | |||
AK044290.1 | - | 3, 4 | 575 | |||
BC039225.1 | - | 3, 4 | 558 | 4458085 |