Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Magee2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053206.2 | NP_444436.1 | 1 | 900 | ||
GenBank mRNA | AF319983.1 | - | 1 | 97 | ||
AK036343.1 | - | 1 | 901 | |||
AK134670.1 | - | 1 | 893 | |||
BC094361.1 | - | 1 | 789 | 6829915 | ||
BC132640.1 | - | 1 | 755 | 40130917 | ||
BC138210.1 | - | 1 | 755 | 8861229 |