Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D11Wsu47e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177777.5 | NP_808445.2 | 2 | 1055 | ||
GenBank mRNA | AK017508.1 | - | 2 | 921 | ||
AK139389.1 | - | 3 | 1011 | |||
AK171307.1 | - | 2 | 964 | |||
AK195222.1 | - | 1 | 90 | |||
BC026908.1 | - | 1 | 668 | 3988907 | ||
BC048942.1 | - | 2 | 1058 | 6442008 |