Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178759.4 | NP_848874.3 | 4 | 702 | ||
GenBank mRNA | AK046560.1 | - | 4 | 701 | ||
AK079787.1 | - | 4 | 701 | |||
AK080042.1 | - | 4 | 701 | |||
AK080212.1 | - | 4 | 703 | |||
AY376716.1 | - | 4 | 701 | |||
AY376717.1 | - | 4 | 701 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC032879.1 | - | 1211 | Chr. - - [-] on Build |