Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wfdc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080550.2 | NP_001074019.1 | 4 | 762 | ||
NM_001276431.1 | NP_001263360.1 | 4 | 762 | |||
NM_001276432.1 | NP_001263361.1 | 4 | 1591 | |||
GenBank mRNA | AK136599.1 | - | 4 | 1592 | ||
AY669506.1 | - | 4 | 762 | |||
AY757956.1 | - | 4 | 762 |