Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc15a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177787.4 | NP_808455.2 | 6 | 1207 | ||
GenBank mRNA | AK036413.1 | - | 5 | 990 | ||
BC026899.1 | - | 3 | 285 | |||
BC039761.1 | - | 3 | 296 | |||
BC141332.1 | - | 5 | 926 | 9055944 | ||
BC141334.1 | - | 5 | 926 | 9055946 |