Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Duxbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183389.1 | NP_899245.1 | 4 | 817 | ||
GenBank mRNA | AK134879.1 | - | 986 | |||
AK141686.1 | - | 4 | 493 | |||
BC055782.1 | - | 4 | 817 | 6403934 | ||
EF472598.1 | - | 4 | 554 |
Gene Symbol: Duxbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177538.1 | NP_001171009.1 | 4 | 807 | ||
GenBank mRNA | EU257807.1 | - | 4 | 391 |
Gene Symbol: Duxbl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177539.1 | NP_001171010.1 | 807 |