Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stkld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198628.2 | NP_941030.2 | 13 | 1225 | ||
GenBank mRNA | BC050806.1 | - | 13 | 1226 |