Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301173.1 | NP_001288102.1 | 5 | 780 | ||
NM_177791.3 | NP_808459.1 | 5 | 780 | |||
GenBank mRNA | AK076644.1 | - | 5 | 772 | ||
BC092530.1 | - | 5 | 780 | 6744085 | ||
BC125431.1 | - | 5 | 772 | 40129946 | ||
BC125433.1 | - | 5 | 772 | 40129948 |