Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175669.3 | NP_783600.1 | 4 | 903 | ||
GenBank mRNA | AK033761.1 | - | 4 | 905 | ||
AY255571.1 | - | 1 | 238 | |||
BC116704.1 | - | 4 | 898 | 40104936 |
Set Membership: |
G-protein coupled receptor |