Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002830.3 | NP_002821.1 | 15 | 2065 | ||
XM_011511560.2 | XP_011509862.1 | 15 | 1970 | |||
XM_011511562.2 | XP_011509864.1 | 14 | 1904 | |||
XM_011511563.2 | XP_011509865.1 | 5 | 1428 | |||
XM_017004596.1 | XP_016860085.1 | 15 | 1991 | |||
XM_017004597.1 | XP_016860086.1 | 15 | 1497 | |||
XM_017004598.1 | XP_016860087.1 | 15 | 1970 | |||
XM_017004599.1 | XP_016860088.1 | 15 | 1476 | |||
XM_017004600.1 | XP_016860089.1 | 15 | 1455 | |||
XM_017004601.1 | XP_016860090.1 | 14 | 1946 | |||
XM_017004602.1 | XP_016860091.1 | 14 | 1925 | |||
XM_017004603.1 | XP_016860092.1 | 14 | 1410 | |||
XM_017004604.1 | XP_016860093.1 | 14 | 1880 | |||
XM_017004605.1 | XP_016860094.1 | 14 | 1776 | |||
XM_017004606.1 | XP_016860095.1 | 14 | 1901 | |||
XM_017004607.1 | XP_016860096.1 | 12 | 1662 | |||
XM_017004608.1 | XP_016860097.1 | 12 | 1619 | |||
GenBank mRNA | AK301218.1 | - | 3 | 281 | ||
AK314836.1 | - | 15 | 1877 | |||
BC010674.1 | - | 15 | 1721 | 3853914 | ||
BX648614.1 | - | 9 | 2299 | |||
M68941.1 | - | 15 | 2065 |