Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrcam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146031.1 | NP_001139503.1 | 19 | 2705 | ||
NM_176930.4 | NP_795904.3 | 20 | 2735 | |||
GenBank mRNA | AJ543321.1 | - | 20 | 2637 | ||
AK087693.1 | - | 19 | 2619 | |||
AK122252.1 | - | 19 | 2635 | |||
AK193959.1 | - | 3 | 336 | |||
AK195700.1 | - | 3 | 339 | |||
AK197224.1 | - | 4 | 364 | |||
AK217961.1 | - | 3 | 337 | |||
AK218614.1 | - | 3 | 338 | |||
BC055053.1 | - | 19 | 2350 |
Set Membership: |
G-protein coupled receptor |