Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176940.5 | NP_795914.3 | 13 | 3423 | ||
GenBank mRNA | AK034868.1 | - | 3122 | |||
AK038792.1 | - | 12 | 3116 | |||
BC082552.1 | - | 13 | 3363 | |||
BC144786.1 | - | 11 | 3013 | 9053334 | ||
BC144787.1 | - | 11 | 3013 | |||
BC145703.1 | - | 11 | 3013 | 40131006 |