Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syne2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005510.2 | NP_001005510.2 | 96, 97 | 17722 | ||
GenBank mRNA | AB093279.1 | - | 12, 13 | 1839 | ||
BC042604.1 | - | 3, 4 | 559 | |||
BC055031.1 | - | 1, 2 | 89 | |||
BC076568.1 | - | 21, 22 | 3296 | |||
BC082586.1 | - | 16, 17 | 2538 |