Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Celf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176954.3 | NP_795928.2 | 11, 12 | 1268 | ||
GenBank mRNA | AK029796.1 | - | 11, 12 | 2442 | ||
AK132321.1 | - | 11, 12 | 1234 | |||
AK158669.1 | - | 10, 11 | 1045 | |||
BC079845.1 | - | 11, 12 | 1239 |