Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPN14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005401.4 | NP_005392.2 | 17 | 3783 | ||
XM_017001941.1 | XP_016857430.1 | 18 | 3870 | |||
GenBank mRNA | AK291641.1 | - | 17 | 3715 | ||
BC101754.1 | - | 16 | 3190 | 8069260 | ||
BC104803.1 | - | 16 | 3190 | 8143806 | ||
HQ116786.1 | - | 15 | 3042 | |||
X82676.1 | - | 17 | 3324 |