Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frmd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028127.3 | NP_082403.2 | 12 | 1643 | ||
GenBank mRNA | AK029709.1 | - | 11 | 1402 | ||
AK054457.1 | - | 12 | 2004 | |||
AK203150.1 | - | 2 | 84 | |||
BC053929.1 | - | 12 | 1643 | 6389695 |