Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp865 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033383.2 | NP_001028555.1 | 2 | 934 | ||
NM_001290426.1 | NP_001277355.1 | 2 | 934 | |||
GenBank mRNA | AK032361.1 | - | 1 | 320 | ||
AK051004.1 | - | 2 | 740 | |||
AK154737.1 | - | 2 | 935 | |||
BC020373.1 | - | 2 | 725 |