Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp865 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033383.2 | NP_001028555.1 | 2 | 2349 | ||
NM_001290426.1 | NP_001277355.1 | 2 | 2349 | |||
GenBank mRNA | AK031793.1 | - | 1 | 1336 | ||
AK032361.1 | - | 1 | 1735 | |||
AK051004.1 | - | 2 | 2155 | |||
AK154737.1 | - | 2 | 2350 | |||
BC115955.1 | - | 1 | 1030 |