Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176996.4 | NP_795970.3 | 9 | 2033 | ||
GenBank mRNA | AK081050.1 | - | 3 | 221 | ||
BC030377.1 | - | 2 | 70 | |||
BC096028.1 | - | 9 | 1552 | 6850065 | ||
BQ947565.1 | - | 4 | 295 | |||
CA749812.1 | - | 6 | 633 | |||
FJ883552.1 | - | 9 | 1542 |
Set Membership: |
G-protein coupled receptor |