Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109754.2 | NP_001103224.1 | 13 | 3167 | ||
NM_001206971.1 | NP_001193900.1 | 10 | 2228 | |||
NM_001206972.1 | NP_001193901.1 | 11 | 2498 | |||
NM_002837.4 | NP_002828.3 | 11 | 2498 | |||
XM_006719528.1 | XP_006719591.1 | 13 | 3167 | |||
XM_006719529.3 | XP_006719592.1 | 11 | 2994 | |||
XM_011538614.1 | XP_011536916.1 | 12 | 2903 | |||
XM_017019723.1 | XP_016875212.1 | 12 | 2903 | |||
XM_017019724.1 | XP_016875213.1 | 5 | 942 | |||
GenBank mRNA | AK128562.1 | - | 13 | 2775 | ||
BC101679.1 | - | 11 | 2497 | 8069185 | ||
BC113463.1 | - | 11 | 2497 | 8322515 | ||
BC143356.1 | - | 11 | 2497 | 9051861 | ||
BC143360.1 | - | 10 | 2227 | 9051865 | ||
BX647238.1 | - | 13 | 3146 | |||
BX648245.1 | - | 13 | 3150 | |||
BX648771.1 | - | 10 | 2346 | |||
CR749546.1 | - | 12 | 2883 | |||
X54131.1 | - | 11 | 2498 |