Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177002.3 | NP_795976.1 | 7 | 1324 | ||
GenBank mRNA | AK040781.1 | - | 7 | 1327 | ||
AK050438.1 | - | 7 | 1326 | |||
AK052592.1 | - | 8 | 1422 | |||
AK080180.1 | - | 7 | 1326 | |||
BC016496.1 | - | 7 | 1278 | |||
BC050265.1 | - | 7 | 1296 | 5044850 | ||
BC132146.1 | - | 7 | 1262 | 40130423 | ||
BC132336.1 | - | 7 | 1262 | 40130613 |
Gene Symbol: Slc22a28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013820.3 | NP_001013842.2 | 7 | 1330 | ||
GenBank mRNA | AK052719.1 | - | 7 | 1331 | ||
BC139174.1 | - | 7 | 1262 | 8862196 |