Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PTPRD SIRNA Assays ›
Gene Symbol:
PTPRD
Gene Name:
protein tyrosine phosphatase, receptor type D
Gene Aliases:
HPTP, HPTPD, HPTPDELTA, PTPD, RPTPDELTA
Chromosome Location:
Chr.9: 8314246 - 10612840 on Build GRCh38
UniGene ID:
Hs.446083
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s11541
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PTPRD
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001040712.2
NP_001035802.1
25
3948
NM_001171025.1
NP_001164496.1
24
3957
NM_002839.3
NP_002830.1
41
5818
NM_130391.3
NP_569075.2
26
3957
NM_130392.3
NP_569076.2
26
3960
NM_130393.3
NP_569077.2
24
3930
XM_006716817.3
XP_006716880.1
42
6167
XM_006716823.2
XP_006716886.1
40
6113
XM_006716825.3
XP_006716888.1
40
6112
XM_006716827.3
XP_006716890.1
37
6052
XM_006716832.3
XP_006716895.1
34
4883
XM_006716833.3
XP_006716896.1
36
4885
XM_006716834.3
XP_006716897.1
34
4886
XM_006716835.3
XP_006716898.1
35
4876
XM_006716837.3
XP_006716900.1
34
4856
XM_006716838.3
XP_006716901.1
34
4855
XM_006716839.3
XP_006716902.1
33
4841
XM_011517992.2
XP_011516294.1
40
6109
XM_017014958.1
XP_016870447.1
42
6175
XM_017014959.1
XP_016870448.1
43
6247
XM_017014960.1
XP_016870449.1
41
6136
XM_017014961.1
XP_016870450.1
41
6120
XM_017014962.1
XP_016870451.1
33
5308
XM_017014963.1
XP_016870452.1
42
6172
XM_017014964.1
XP_016870453.1
42
6171
XM_017014965.1
XP_016870454.1
42
6174
XM_017014966.1
XP_016870455.1
41
6166
XM_017014967.1
XP_016870456.1
41
6161
XM_017014968.1
XP_016870457.1
41
6157
XM_017014969.1
XP_016870458.1
41
6157
XM_017014970.1
XP_016870459.1
41
6157
XM_017014971.1
XP_016870460.1
40
6146
XM_017014972.1
XP_016870461.1
41
6148
XM_017014973.1
XP_016870462.1
40
6142
XM_017014974.1
XP_016870463.1
41
6145
XM_017014975.1
XP_016870464.1
39
6131
XM_017014976.1
XP_016870465.1
41
6130
XM_017014977.1
XP_016870466.1
40
6131
XM_017014978.1
XP_016870467.1
41
6127
XM_017014979.1
XP_016870468.1
41
6126
XM_017014980.1
XP_016870469.1
39
6082
XM_017014981.1
XP_016870470.1
36
6038
XM_017014982.1
XP_016870471.1
37
4938
XM_017014983.1
XP_016870472.1
37
4935
XM_017014984.1
XP_016870473.1
36
4926
XM_017014985.1
XP_016870474.1
36
4920
XM_017014986.1
XP_016870475.1
36
4916
XM_017014987.1
XP_016870476.1
35
4901
XM_017014988.1
XP_016870477.1
35
4898
XM_017014989.1
XP_016870478.1
36
4892
XM_017014990.1
XP_016870479.1
36
4888
XM_017014991.1
XP_016870480.1
36
4888
XM_017014992.1
XP_016870481.1
35
4879
XM_017014993.1
XP_016870482.1
35
4874
XM_017014994.1
XP_016870483.1
35
4871
XM_017014995.1
XP_016870484.1
35
4877
GenBank mRNA
AB208869.1
-
24
3774
AB211400.1
-
40
5786
AK296392.1
-
9
1215
BC106713.1
-
24
3905
40027581
BC106714.1
-
26
3908
40027582
BC106715.2
-
26
3905
40027584
BC106716.1
-
24
3878
40027585
L38929.1
-
31
5228
X54133.1
-
21
3908
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein dephosphorylation
phosphate-containing compound metabolic process
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
transmembrane receptor protein tyrosine phosphatase signaling pathway
chemical synaptic transmission
neuron differentiation
peptidyl-tyrosine dephosphorylation
positive regulation of dendrite morphogenesis
presynaptic membrane assembly
transmembrane receptor protein tyrosine phosphatase activity
receptor binding
protein binding
cell adhesion molecule binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-8tsm6:80/100.66.77.47:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline