Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cobll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027225.1 | NP_081501.1 | 12 | 3605 | ||
NM_177025.5 | NP_795999.2 | 13 | 3716 | |||
GenBank mRNA | AK037010.1 | - | 14 | 3710 | ||
AK052533.1 | - | 12 | 3554 | |||
AK135859.1 | - | 4 | 1944 | |||
AK144943.1 | - | 14 | 3763 | |||
AK173076.1 | - | 12 | 3436 | |||
AY308746.1 | - | 13 | 3716 | |||
BC067007.1 | - | 12 | 3604 | 5702930 |