Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dsel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081316.1 | NP_001074785.1 | 2 | 3397 | ||
GenBank mRNA | AK032110.1 | - | 1 | 1288 | ||
AK033983.1 | - | 2 | 3398 | |||
BC120567.1 | - | 1 | 1759 | 40129490 | ||
BC137796.1 | - | 1 | 1759 | 8860815 |